Merck
  • Targeted Phage Display-based Pulmonary Vaccination in Mice and Non-human Primates.
Targeted Phage Display-based Pulmonary Vaccination in Mice and Non-human Primates.
Med (New York, N.Y.) (2021-04-20)
Daniela I Staquicini, E Magda Barbu, Rachel L Zemans, Beth K Dray, Fernanda I Staquicini, Prashant Dogra, Marina Cardó-Vila, Cindy K Miranti, Wallace B Baze, Luisa L Villa, Jorge Kalil, Geetanjali Sharma, Eric R Prossnitz, Zhihui Wang, Vittorio Cristini, Richard L Sidman, Andrew R Berman, Reynold A Panettieri, Rubin M Tuder, Renata Pasqualini, Wadih Arap
ABSTRACT
The extensive alveolar capillary network of the lungs is an attractive route for administration of several agents. One key functional attribute is the rapid onset of systemic action due to the absence of first-pass metabolism. Here we applied a combinatorial approach for ligand-directed pulmonary delivery as a unique route for systemic targeting in vaccination. We screened a phage display random peptide library in vivo to select, identify, and validate a ligand (CAKSMGDIVC) that specifically targets and is internalized through its receptor, α3β1 integrin, on the surface of cells lining the lung airways and alveoli and mediates CAKSMGDIVC-displaying phage binding and systemic delivery without compromising lung homeostasis. As a proof-of-concept, we show that the pulmonary delivery of targeted CAKSMGDIVC-displaying phage particles in mice and non-human primates elicit a systemic and specific humoral response. This broad methodology blueprint represents a robust and versatile platform tool enabling new ligand-receptor discovery with many potential translational applications. Cancer Center Support Grants to the University of Texas M.D. Anderson Cancer Center (CA016672), University of New Mexico Comprehensive Cancer Center (CA118100), Rutgers Cancer Institute of New Jersey (CA072720), research awards from the Gillson Longenbaugh Foundation, and National Institutes of Health (NIH) grant no. 1R01CA226537.
Sigma-Aldrich
Lipopolysaccharides from Klebsiella pneumoniae, purified by phenol extraction
Sigma-Aldrich
Anti-Clara Cell Secretory Protein Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Prosurfactant Protein C (proSP-C) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-fd Bacteriophage antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone ASC-1 (Azide Free), clone ASC-1, Chemicon®, from mouse