Merck
  • Radiative lifetime-encoded unicolour security tags using perovskite nanocrystals.
Radiative lifetime-encoded unicolour security tags using perovskite nanocrystals.
Nature communications (2021-02-14)
Sergii Yakunin, Jana Chaaban, Bogdan M Benin, Ihor Cherniukh, Caterina Bernasconi, Annelies Landuyt, Yevhen Shynkarenko, Sami Bolat, Christoph Hofer, Yaroslav E Romanyuk, Stefano Cattaneo, Sergey I Pokutnyi, Richard D Schaller, Maryna I Bodnarchuk, Dimos Poulikakos, Maksym V Kovalenko
ABSTRACT
Traditional fluorescence-based tags, used for anticounterfeiting, rely on primitive pattern matching and visual identification; additional covert security features such as fluorescent lifetime or pattern masking are advantageous if fraud is to be deterred. Herein, we present an electrohydrodynamically printed unicolour multi-fluorescent-lifetime security tag system composed of lifetime-tunable lead-halide perovskite nanocrystals that can be deciphered with both existing time-correlated single-photon counting fluorescence-lifetime imaging microscopy and a novel time-of-flight prototype. We find that unicolour or matching emission wavelength materials can be prepared through cation-engineering with the partial substitution of formamidinium for ethylenediammonium to generate "hollow" formamidinium lead bromide perovskite nanocrystals; these materials can be successfully printed into fluorescence-lifetime-encoded-quick-read tags that are protected from conventional readers. Furthermore, we also demonstrate that a portable, cost-effective time-of-flight fluorescence-lifetime imaging prototype can also decipher these codes. A single comprehensive approach combining these innovations may be eventually deployed to protect both producers and consumers.
Sigma-Aldrich
Didecyldimethylammonium bromide, 98%
Sigma-Aldrich
Ethylenediamine diacetate, 98%
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Formamidine acetate salt, 99%
Sigma-Aldrich
Methyl acetate, suitable for HPLC, ≥99.8%