- Entry of spores into intestinal epithelial cells contributes to recurrence of Clostridioides difficile infection.
Entry of spores into intestinal epithelial cells contributes to recurrence of Clostridioides difficile infection.
Nature communications (2021-02-20)
Pablo Castro-Córdova, Paola Mora-Uribe, Rodrigo Reyes-Ramírez, Glenda Cofré-Araneda, Josué Orozco-Aguilar, Christian Brito-Silva, María José Mendoza-León, Sarah A Kuehne, Nigel P Minton, Marjorie Pizarro-Guajardo, Daniel Paredes-Sabja
PMID33602902
ABSTRACT
Clostridioides difficile spores produced during infection are important for the recurrence of the disease. Here, we show that C. difficile spores gain entry into the intestinal mucosa via pathways dependent on host fibronectin-α5β1 and vitronectin-αvβ1. The exosporium protein BclA3, on the spore surface, is required for both entry pathways. Deletion of the bclA3 gene in C. difficile, or pharmacological inhibition of endocytosis using nystatin, leads to reduced entry into the intestinal mucosa and reduced recurrence of the disease in a mouse model. Our findings indicate that C. difficile spore entry into the intestinal barrier can contribute to spore persistence and infection recurrence, and suggest potential avenues for new therapies.
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone B3A, azide free, clone B3A, Chemicon®, from mouse
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P5D2 (Azide Free)| MAB1959, clone P5D2, Chemicon®, from mouse
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse