Merck
Structure and Function of the Bacterial Protein Toxin Phenomycin.
Structure (London, England : 1993) (2020-03-30)
Bente K Hansen, Camilla K Larsen, Jakob T Nielsen, Esben B Svenningsen, Lan B Van, Kristian M Jacobsen, Morten Bjerring, Rasmus K Flygaard, Lasse B Jenner, Lene N Nejsum, Ditlev E Brodersen, Frans A A Mulder, Thomas Tørring, Thomas B Poulsen
ABSTRACT
Phenomycin is a bacterial mini-protein of 89 amino acids discovered more than 50 years ago with toxicity in the nanomolar regime toward mammalian cells. The protein inhibits the function of the eukaryotic ribosome in cell-free systems and appears to target translation initiation. Several fundamental questions concerning the cellular activity of phenomycin, however, have remained unanswered. In this paper, we have used morphological profiling to show that direct inhibition of translation underlies the toxicity of phenomycin in cells. We have performed studies of the cellular uptake mechanism of phenomycin, showing that endosomal escape is the toxicity-limiting step, and we have solved a solution phase high-resolution structure of the protein using NMR spectroscopy. Through bioinformatic as well as functional comparisons between phenomycin and two homologs, we have identified a peptide segment, which constitutes one of two loops in the structure that is critical for the toxicity of phenomycin.
Sigma-Aldrich
Ala-Gln, 200 mM, solution, sterile-filtered, Biotechnology Performance Certified
Sigma-Aldrich
McCoy′s 5A Medium, Modified, with L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt >300,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%