Merck
  • Influenza A viral replication is blocked by inhibition of the inositol-requiring enzyme 1 (IRE1) stress pathway.
Influenza A viral replication is blocked by inhibition of the inositol-requiring enzyme 1 (IRE1) stress pathway.
The Journal of biological chemistry (2011-12-24)
Ihab H Hassan, Michael S Zhang, Linda S Powers, Jian Q Shao, Jonas Baltrusaitis, D Thomas Rutkowski, Kevin Legge, Martha M Monick
ABSTRACT
Known therapies for influenza A virus infection are complicated by the frequent emergence of resistance. A therapeutic strategy that may escape viral resistance is targeting host cellular mechanisms involved in viral replication and pathogenesis. The endoplasmic reticulum (ER) stress response, also known as the unfolded protein response (UPR), is a primitive, evolutionary conserved molecular signaling cascade that has been implicated in multiple biological phenomena including innate immunity and the pathogenesis of certain viral infections. We investigated the effect of influenza A viral infection on ER stress pathways in lung epithelial cells. Influenza A virus induced ER stress in a pathway-specific manner. We showed that the virus activates the IRE1 pathway with little or no concomitant activation of the PERK and the ATF6 pathways. When we examined the effects of modulating the ER stress response on the virus, we found that the molecular chaperone tauroursodeoxycholic acid (TUDCA) significantly inhibits influenza A viral replication. In addition, a specific inhibitor of the IRE1 pathway also blocked viral replication. Our findings constitute the first evidence that ER stress plays a role in the pathogenesis of influenza A viral infection. Decreasing viral replication by modulating the host ER stress response is a novel strategy that has important therapeutic implications.
Product Description
Millipore
Phosphatase Inhibitor Cocktail Set II, A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.
Millipore
Tauroursodeoxycholic Acid, Sodium Salt, Tauroursodeoxycholic is a detergent useful for the solubilization of lipids and membrane-bound proteins.
Sigma-Aldrich
PhosphoDetect Anti-SAPK/JNK (pThr¹⁸³, Tyr¹⁸⁵) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Tunicamycin, Streptomyces lysosuperficus