Merck
  • CDK5RAP3 as tumour suppressor negatively regulates self-renewal and invasion and is regulated by ERK1/2 signalling in human gastric cancer.
CDK5RAP3 as tumour suppressor negatively regulates self-renewal and invasion and is regulated by ERK1/2 signalling in human gastric cancer.
British journal of cancer (2020-07-02)
Jian-Xian Lin, Changhwan Yoon, Ping Li, Sandra W Ryeom, Soo-Jeong Cho, Chao-Hui Zheng, Jian-Wei Xie, Jian-Bin Wang, Jun Lu, Qi-Yue Chen, Sam S Yoon, Chang-Ming Huang
ABSTRACT
Toward identifying new strategies to target gastric cancer stem-like cells (CSCs), we evaluated the function of the tumour suppressor CDK5 regulatory subunit-associated protein 3 (CDK5RAP3) in gastric CSC maintenance. We examined the expression of CDK5RAP3 and CD44 in gastric cancer patients. The function and mechanisms of CDK5RAP3 were checked in human and mouse gastric cancer cell lines and in mouse xenograft. We show that CDK5RAP3 is weakly expressed in gastric CSCs and is negatively correlated with the gastric CSC marker CD44. CDK5RAP3 overexpression decreased expression of CSC markers, spheroid formation, invasion and migration, and reversed chemoresistance in gastric CSCs in vitro and vivo. CDK5RAP3 expression was found to be regulated by extracellular-related kinase (ERK) signalling. ERK inhibitors decreased spheroid formation, migration and invasion, and the expression of epithelial-to-mesenchymal transition (EMT)-related proteins in both GA cells and organoids derived from a genetically engineered mouse model of GA. Finally, CDK5RAP3 expression was associated with reduced lymph-node metastasis and better prognosis, even in the presence of high expression of the EMT transcription factor Snail, among patients with CD44-positive GA. Our results demonstrate that CDK5RAP3 is suppressed by ERK signalling and negatively regulates the self-renewal and EMT of gastric CSCs.
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK5RAP3
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Millipore
SB 202190, SB 202190, CAS 152121-30-7, is a potent, reversible, competitive inhibitor of p38. Inhibits p38 phosphorylation of myelin basic protein. Blocks the activity of p38β (Ki = 16 nM; IC₅₀ = 350 nM).
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).