Merck
Redox homeostasis maintained by GPX4 facilitates STING activation.
Nature immunology (2020-06-17)
Mutian Jia, Danhui Qin, Chunyuan Zhao, Li Chai, Zhongxia Yu, Wenwen Wang, Li Tong, Lin Lv, Yuanyuan Wang, Jan Rehwinkel, Jinming Yu, Wei Zhao
ABSTRACT
Stimulator-of-interferon genes (STING) is vital for sensing cytosolic DNA and initiating innate immune responses against microbial infection and tumors. Redox homeostasis is the balance of oxidative and reducing reactions present in all living systems. Yet, how the intracellular redox state controls STING activation is unclear. Here, we show that cellular redox homeostasis maintained by glutathione peroxidase 4 (GPX4) is required for STING activation. GPX4 deficiency enhanced cellular lipid peroxidation and thus specifically inhibited the cGAS-STING pathway. Concordantly, GPX4 deficiency inhibited herpes simplex virus-1 (HSV-1)-induced innate antiviral immune responses and promoted HSV-1 replication in vivo. Mechanistically, GPX4 inactivation increased production of lipid peroxidation, which led to STING carbonylation at C88 and inhibited its trafficking from the endoplasmic reticulum (ER) to the Golgi complex. Thus, cellular stress-induced lipid peroxidation specifically attenuates the STING DNA-sensing pathway, suggesting that GPX4 facilitates STING activation by maintaining redox homeostasis of lipids.
Sigma-Aldrich
HNE-DMA, hexane solution, ≥85% (GC)
Sigma-Aldrich
DMXAA, ≥98% (HPLC), solid
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Phosphatidylinositol sodium salt, from soybean
Sigma-Aldrich
L-α-Phosphatidylcholine, from soybean, Type IV-S, ≥30% (enzymatic)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
9-Oxo-10(9H)-acridineacetic acid, for HPLC derivatization, ≥99.0% (T)
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
3-Ethynylaniline, ≥98%
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution