Merck
  • Synthesis of molecularly imprinted polymer adsorbents for solid-phase extraction of strobilurin fungicides from agricultural products.
Synthesis of molecularly imprinted polymer adsorbents for solid-phase extraction of strobilurin fungicides from agricultural products.
Journal of separation science (2020-02-27)
Xinyi Li, Xiaolin Cao, Zheng Zhang, Ziping Zhang, Zejun Jiang, Jungang Yin
ABSTRACT
Molecularly imprinted polymers for strobilurin fungicides were prepared by precipitation polymerization employing azoxystrobin as template molecular together with methacrylic acid monomer and trimethylolpropane triacrylate cross-linker. Morphological characterization showed molecularly imprinted polymers were uniform spherical particles with about 0.2 μm in diameter, while the morphologies of nonimprinted polymers were irregular bulk. The equilibrium binding and selective experiments proved that molecularly imprinted polymers possessed a higher affinity toward four fungicides compared to nonimprinted polymers and heterogeneous binding sites were found in the molecularly imprinted polymers. Molecularly imprinted solid-phase extraction conditions, including sample loading solvents, selective washing, and elution solvents, were carefully optimized. The developed method showed good recoveries (70.0-114.0%) with relative standard deviations in range of 1.0-9.8% (n  =  3) for samples (cucumber and peach) spiked at three different levels (10, 50, and 100 μg/ kg). The detection limit (signal/noise = 3) ranged from 0.01 to 0.08 μg/kg. The results demonstrated good potential use of this convenient and highly efficient method for determining trace strobilurin fungicides in agricultural products.
Product Description
Supelco
Picoxystrobin, PESTANAL®, analytical standard
Sigma-Aldrich
Trimethylolpropane triacrylate, contains monomethyl ether hydroquinone as inhibitor, technical grade
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Supelco
Dimoxystrobin, PESTANAL®, analytical standard
Supelco
Azoxystrobin, PESTANAL®, analytical standard