Merck
  • Single-cell analysis of RORα tracer mouse lung reveals ILC progenitors and effector ILC2 subsets.
Single-cell analysis of RORα tracer mouse lung reveals ILC progenitors and effector ILC2 subsets.
The Journal of experimental medicine (2019-12-10)
Maryam Ghaedi, Zi Yi Shen, Mona Orangi, Itziar Martinez-Gonzalez, Lisa Wei, Xiaoxiao Lu, Arundhoti Das, Alireza Heravi-Moussavi, Marco A Marra, Avinash Bhandoola, Fumio Takei
ABSTRACT
Lung group 2 innate lymphoid cells (ILC2s) drive allergic inflammation and promote tissue repair. ILC2 development is dependent on the transcription factor retinoic acid receptor-related orphan receptor (RORα), which is also expressed in common ILC progenitors. To elucidate the developmental pathways of lung ILC2s, we generated RORα lineage tracer mice and performed single-cell RNA sequencing, flow cytometry, and functional analyses. In adult mouse lungs, we found an IL-18Rα+ST2- population different from conventional IL-18Rα-ST2+ ILC2s. The former was GATA-3intTcf7EGFP+Kit+, produced few cytokines, and differentiated into multiple ILC lineages in vivo and in vitro. In neonatal mouse lungs, three ILC populations were identified, namely an ILC progenitor population similar to that in adult lungs and two distinct effector ILC2 subsets that differentially produced type 2 cytokines and amphiregulin. Lung ILC progenitors might actively contribute to ILC-poiesis in neonatal and inflamed adult lungs. In addition, neonatal lung ILC2s include distinct proinflammatory and tissue-repairing subsets.
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Papain from Carica papaya, solution, light brown, ≥10 U/mg protein (~25 mg/ml)