Merck
Fluorination of arylboronic esters enabled by bismuth redox catalysis.
Science (New York, N.Y.) (2020-01-18)
Oriol Planas, Feng Wang, Markus Leutzsch, Josep Cornella
ABSTRACT
Bismuth catalysis has traditionally relied on the Lewis acidic properties of the element in a fixed oxidation state. In this paper, we report a series of bismuth complexes that can undergo oxidative addition, reductive elimination, and transmetallation in a manner akin to transition metals. Rational ligand optimization featuring a sulfoximine moiety produced an active catalyst for the fluorination of aryl boronic esters through a bismuth (III)/bismuth (V) redox cycle. Crystallographic characterization of the different bismuth species involved, together with a mechanistic investigation of the carbon-fluorine bond-forming event, identified the crucial features that were combined to implement the full catalytic cycle.
Sigma-Aldrich
1-Chloro-3-fluorobenzene, 99%
Sigma-Aldrich
3-Fluoroanisole, 99%
Sigma-Aldrich
4-Fluorobenzonitrile, 99%
Sigma-Aldrich
2,6-Dichloro-1-fluoropyridinium tetrafluoroborate, 97%