Merck
Vasculogenic mimicry in small cell lung cancer.
Nature communications (2016-11-09)
Stuart C Williamson, Robert L Metcalf, Francesca Trapani, Sumitra Mohan, Jenny Antonello, Benjamin Abbott, Hui Sun Leong, Christopher P E Chester, Nicole Simms, Radoslaw Polanski, Daisuke Nonaka, Lynsey Priest, Alberto Fusi, Fredrika Carlsson, Anders Carlsson, Mary J C Hendrix, Richard E B Seftor, Elisabeth A Seftor, Dominic G Rothwell, Andrew Hughes, James Hicks, Crispin Miller, Peter Kuhn, Ged Brady, Kathryn L Simpson, Fiona H Blackhall, Caroline Dive
ABSTRACT
Small cell lung cancer (SCLC) is characterized by prevalent circulating tumour cells (CTCs), early metastasis and poor prognosis. We show that SCLC patients (37/38) have rare CTC subpopulations co-expressing vascular endothelial-cadherin (VE-cadherin) and cytokeratins consistent with vasculogenic mimicry (VM), a process whereby tumour cells form 'endothelial-like' vessels. Single-cell genomic analysis reveals characteristic SCLC genomic changes in both VE-cadherin-positive and -negative CTCs. Higher levels of VM are associated with worse overall survival in 41 limited-stage patients' biopsies (P<0.025). VM vessels are also observed in 9/10 CTC patient-derived explants (CDX), where molecular analysis of fractionated VE-cadherin-positive cells uncovered copy-number alterations and mutated TP53, confirming human tumour origin. VE-cadherin is required for VM in NCI-H446 SCLC xenografts, where VM decreases tumour latency and, despite increased cisplatin intra-tumour delivery, decreases cisplatin efficacy. The functional significance of VM in SCLC suggests VM regulation may provide new targets for therapeutic intervention.
Sigma-Aldrich
GenomePlex® Single Cell Whole Genome Amplification Kit, Amplify genome of a single cell
Sigma-Aldrich
Anti-Cisplatin DNA Adducts Antibody, clone ICR4, clone 1CR4, from rat