Merck
  • RBX2 maintains final retinal cell position in a DAB1-dependent and -independent fashion.
RBX2 maintains final retinal cell position in a DAB1-dependent and -independent fashion.
Development (Cambridge, England) (2018-01-24)
Corinne L Fairchild, Keiko Hino, Jisoo S Han, Adam M Miltner, Gabriel Peinado Allina, Caileigh E Brown, Marie E Burns, Anna La Torre, Sergi Simó
ABSTRACT
The laminated structure of the retina is fundamental for the organization of the synaptic circuitry that translates light input into patterns of action potentials. However, the molecular mechanisms underlying cell migration and layering of the retina are poorly understood. Here, we show that RBX2, a core component of the E3 ubiquitin ligase CRL5, is essential for retinal layering and function. RBX2 regulates the final cell position of rod bipolar cells, cone photoreceptors and Muller glia. Our data indicate that sustained RELN/DAB1 signaling, triggered by depletion of RBX2 or SOCS7 - a CRL5 substrate adaptor known to recruit DAB1 - causes rod bipolar cell misposition. Moreover, whereas SOCS7 also controls Muller glia cell lamination, it is not responsible for cone photoreceptor positioning, suggesting that RBX2, most likely through CRL5 activity, controls other signaling pathways required for proper cone localization. Furthermore, RBX2 depletion reduces the number of ribbon synapses and disrupts cone photoreceptor function. Together, these results uncover RBX2 as a crucial molecular regulator of retina morphogenesis and cone photoreceptor function.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-DAB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Protein Kinase C (PKC) antibody produced in mouse, clone MC5, ascites fluid
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Cone Arrestin Antibody, Chemicon®, from rabbit