Merck
  • Rising above helium: A hydrogen carrier gas chromatography flame ionization detection (GC-FID) method for the simultaneous quantification of toxic alcohols and ethylene glycol in human plasma specimens.
Rising above helium: A hydrogen carrier gas chromatography flame ionization detection (GC-FID) method for the simultaneous quantification of toxic alcohols and ethylene glycol in human plasma specimens.
Clinical biochemistry (2019-08-20)
Joshua Buse, Jason L Robinson, Rosemary Shyne, Qingli Chi, Darlene Affleck, Donna Duce, Isolde Seiden-Long
ABSTRACT
Here we validate a GC, Flame Ionization Detection (GC-FID), liquid injection method using hydrogen as a carrier gas combining analysis of toxic volatile alcohols (VA): methanol, ethanol, isopropanol, acetone, as well as glycols, ethylene glycol (EG) and propylene glycol (PG), in a single method. 200 μL of calibrator, QC, or patient specimen were deproteinized with 400 μL of acetonitrile containing internal standards (10 mmol/L N-propyl alcohol for VA and 2.5 mmol/L 1,2-butanediol for glycols). GC-FID analysis using hydrogen carrier gas and nitrogen makeup gas utilized an Agilent 7890 system equipped with Agilent 7683 liquid autosampler on a 30 m × 530 μm RTX-200 fused silica column. Method validation included repeatability, recovery, carryover, linearity, lower limit of quantification (LLOQ), accuracy, selectivity and measurement uncertainty. The 8.3 min from injection to injection reduced time of analysis by 45% over a previously reported method using Helium carrier gas with no loss in resolution. Within-run and Between-run variability were ≤1.4% and ≤6.8% respectively. Recovery was 100% within a 95% confidence interval. Carryover was negligible for all but EG. LLOQ was <1 mmol/L for all analytes. The upper range of linearity was 120 mmol/L for methanol, ethanol and isopropanol, 100 mmol/L for acetone and 50 mmol/L for EG. Analytes demonstrated acceptable accuracy and measurement uncertainty using College of American Pathologists (CAP) criteria. Toluene can cause a false positive EG, while benzene, xylene and 1,3 butanediol can cause false negative EG. Converting from Helium to Hydrogen carrier gas benefits patient care through a reduction in turnaround time and provides a cost savings to the laboratory.
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Butanediol, 98%
Sigma-Aldrich
Intralipid, 20%, emulsion