Merck
  • The thrombospondin motif of aggrecanase-1 (ADAMTS-4) is critical for aggrecan substrate recognition and cleavage.
The thrombospondin motif of aggrecanase-1 (ADAMTS-4) is critical for aggrecan substrate recognition and cleavage.
The Journal of biological chemistry (2000-05-29)
M Tortorella, M Pratta, R Q Liu, I Abbaszade, H Ross, T Burn, E Arner
ABSTRACT
Aggrecanase-1 (ADAMTS-4) is a member of the a disintegrin and metalloprotease with thrombospondin motifs (ADAMTS) protein family that was recently identified. Aggrecanase-1 is one of two ADAMTS cartilage-degrading enzymes purified from interleukin-1-stimulated bovine nasal cartilage (Tortorella, M. D., Burn, T. C., Pratta, M. A. , Abbaszade, I., Hollis, J. M., Liu, R., Rosenfeld, S. A., Copeland, R. A., Decicco, C. P., Wynn, R., Rockwell, A., Yang, F., Duke, J. L., Solomon, K., George, H., Bruckner, R., Nagase, H., Itoh, Y., Ellis, D. M., Ross, H., Wiswall, B. H., Murphy, K., Hillman, M. C., Jr., Hollis, G. F., and Arner, E.C. (1999) Science 284, 1664-1666; 2 Abbaszade, I., Liu, R. Q., Yang, F., Rosenfeld, S. A., Ross, O. H., Link, J. R., Ellis, D. M., Tortorella, M. D., Pratta, M. A., Hollis, J. M., Wynn, R., Duke, J. L., George, H. J., Hillman, M. C., Jr., Murphy, K., Wiswall, B. H., Copeland, R. A., Decicco, C. P., Bruckner, R., Nagase, H., Itoh, Y., Newton, R. C., Magolda, R. L., Trzaskos, J. M., and Burn, T. C. (1999) J. Biol. Chem. 274, 23443-23450). The aggrecan products generated by this enzyme are found in cartilage cultures stimulated with cytokines and in synovial fluid from patients with arthritis, suggesting that aggrecanase-1 may be important in diseases involving cartilage destruction. Here we demonstrate that the thrombospondin type-1 (TSP-1) motif located within the C terminus of aggrecanase-1 binds to the glycosaminoglycans of aggrecan. Data from several studies indicate that this binding of aggrecanase-1 to aggrecan through the TSP-1 motif is necessary for enzymatic cleavage of aggrecan. 1) A truncated form of aggrecanase-1 lacking the TSP-1 motif was not effective in cleaving aggrecan. 2) Several peptides representing different regions of the TSP-1 motif effectively blocked aggrecanase-1 cleavage of aggrecan by preventing the enzyme from binding to the substrate. 3) Aggrecanase-1 was not effective in cleaving glycosaminoglycan-free aggrecan. Taken together, these data suggest that the TSP-1 motif of aggrecanase-1 is critical for substrate recognition and cleavage.
Sigma-Aldrich
ADAMTS-4, recombinant, His Tagged, This His Tagged Recombinant ADAMTS4 is used to study the degradation of extracellular matrix proteoglycans, to screen for inhibitors of proteoglycan hydrolysis and to characterize inhibitor actions.
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Anti-Chondroitin 6 Sulfate Antibody, clone MK-302, ascites fluid, clone MK-302, Chemicon®
Pricing and availability is not currently available. Contact Technical Service