Merck
  • Synergistic effect of Nutlin-3 combined with MG-132 on schwannoma cells through restoration of merlin and p53 tumour suppressors.
Synergistic effect of Nutlin-3 combined with MG-132 on schwannoma cells through restoration of merlin and p53 tumour suppressors.
EBioMedicine (2018-10-03)
Hongsai Chen, Lu Xue, He Huang, Hantao Wang, Xiaoman Zhang, Weidong Zhu, Zhigang Wang, Zhaoyan Wang, Hao Wu
ABSTRACT
The great majority of sporadic vestibular schwannomas (VSs) are due to the mutations of the NF2 gene encoding merlin. Sporadic VSs exhibit variable growth patterns and only a small fraction of the tumours are fast-growing; however, the underlying mechanisms remain undefined. DNA sequencing and dosage analysis were used to identify the NF2 mutation status in sporadic schwannomas. The expression and sub-cellular localization of merlin and p53-MDM2 were assessed by immunoblotting, qRT-PCR and immunofluorescence. In vitro and in vivo studies were performed to reveal the effects of Nutlin-3 (a MDM2 inhibitor) and/or MG-132(a proteasome inhibitor) on schwannomas. The proliferation of schwannoma cells was assessed by CCK-8 assay, EdU staining and Flow cytometry analysis. Double genetic hits of NF2 tended to occur in fast-growing tumours, characterized by the absence of merlin. The deregulation of p53-MDM2 was demonstrated to mediate merlin-deficient tumour growth, characterized by a nuclear accumulation of stabilized MDM2, contributing to a nuclear export of p53 for degradation. Nutlin-3 blocked the proliferation of schwannoma cells via a cooperative recovery of merlin and p53, accompanied by the shuttling of both proteins from the cytoplasm to the nucleus. We further demonstrated a difference in the sensitivity to Nutlin-3 between schwannoma cells with and without merlin expression. Nutlin-3 combined with MG-132 narrowed this between-group difference and triggered stronger inhibitory effects on the growth of schwannomas through coordinated reactivation of p53. These findings present treatment strategies directed on the pathogenesis of sporadic schwannomas. FUND: National Natural Science Foundation of China.
Sigma-Aldrich
Monoclonal Anti-Cyclin D1 antibody produced in mouse, clone DCS-6, ascites fluid
Sigma-Aldrich
Anti-MDM2 (Ab-166) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-NF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-1, purified from hybridoma cell culture