Merck
The p53 proto-oncogene can act as a suppressor of transformation.
Cell (1989-06-30)
C A Finlay, P W Hinds, A J Levine
ABSTRACT
DNA clones of the wild-type p53 proto-oncogene inhibit the ability of E1A plus ras or mutant p53 plus ras-activated oncogenes to transform primary rat embryo fibroblasts. The rare clones of transformed foci that result from E1A plus ras plus wild-type p53 triple transfections all contain the p53 DNA in their genome, but the great majority fail to express the p53 protein. The three cell lines derived from such foci that express p53 all produce mutant p53 proteins with properties similar or identical to transformation-activated p53 proteins. The p53 mutants selected in this fashion (transformation in vitro) resemble the p53 mutants selected in tumors (in vivo). These results suggest that the p53 proto-oncogene can act negatively to block transformation.
Sigma-Aldrich
p53 (1-81), mutant, GST tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
p53 (1-81), wild type, GST tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)