Skip to Content
Merck
Ytterbium(III) chloride hexahydrate 99.998% trace metals basis

Ytterbium(III) chloride hexahydrate

Ytterbium(III) chloride hexahydrate 99.998% trace metals basis
Synonym(s):
Ytterbium trichloride hydrate, Ytterbium(III) chloride·6hydrate
Linear Formula:
YbCl3 · 6H2O
CAS Number:
Molecular Weight:
387.49
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2