
Valganciclovir containing impurity T

Synonym(s):
Valganciclovir hydrochloride, L-Valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl ester monohydrochloride
Empirical Formula (Hill Notation):
C14H22N6O5 · HCl
CAS Number:
Molecular Weight:
390.82
Products
Brand
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing