
Trospium chloride Related Compound B

Synonym(s):
Nortropinyl benzilate hydrochloride, (1R,3r,5S)-8-Azabicyclo[3.2.1]octan-3-yl hydroxydiphenylacetate hydrochloride
Empirical Formula (Hill Notation):
C21H23NO3 · HCl
CAS Number:
Molecular Weight:
373.87
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2