Skip to Content
Merck
Tris(dimethylamino)phosphine 97%

Tris(dimethylamino)phosphine

Tris(dimethylamino)phosphine 97%
Synonym(s):
HMPT, Hexamethylphosphorous triamide
Linear Formula:
P[N(CH3)2]3
CAS Number:
Molecular Weight:
163.20

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing