
Topiramate Related Compound A

Synonym(s):
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose, D-Fructopyranose diacetonide
Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing