Skip to Content
Merck
Tetramethylammonium fluoride tetrahydrate 98%

Tetramethylammonium fluoride tetrahydrate

Tetramethylammonium fluoride tetrahydrate 98%
Linear Formula:
(CH3)4N(F) · 4H2O
CAS Number:
Molecular Weight:
165.20

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing