Skip to Content
Merck
Testosterone cypionate ≥98% (HPLC)

Testosterone cypionate

Testosterone cypionate ≥98% (HPLC)
Synonym(s):
Testosterone 17β-cypionate, 17β-Hydroxy-4-androsten-3-one 17-cyclopentylpropionate, 4-Androsten-17β-ol-3-one 17-cypionate, Depotestosterone, Depovirin, Testosterone 17β-cyclopentanepropionate, (17β)-17-(3-Cyclopentyl-1-oxopropoxy)androst-4-en-3-one, 17β-Hydroxyandrost-4-en-3-one cyclopentylpropionate, Cyclopentanepropionic acid ester with testosterone, NSC 9157
Empirical Formula (Hill Notation):
C27H40O3
CAS Number:
Molecular Weight:
412.60

Products

Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing