
Terpin impurity D

Synonym(s):
trans-Terpin, 4-(2-Hydroxyisopropyl)-1-methylcyclohexanol, p-Menthane-1,8-diol
Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
Products
Formula Weight
Manufacturer
Melting Point (°C)
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing