Skip to Content
Merck
vibrant-m

Tacrolimus 8-epimer

vibrant-m
Synonym(s):
8-epi-Tacrolimus, Epitacrolimus
Empirical Formula (Hill Notation):
C44H69NO12
CAS Number:
Molecular Weight:
804.02

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade