
Sufentanil citrate

Synonym(s):
N-[4-(Methoxymethyl)-1-[2-(2-thienyl)ethyl]-4-piperidinyl]-N-phenylpropanamide citrate, R-33800, Sufentanyl citrate
Empirical Formula (Hill Notation):
C22H30N2O2S · C6H8O7
CAS Number:
Molecular Weight:
578.67
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing