
Sodium dihydrogen phosphate monohydrate

Synonym(s):
Sodium phosphate monobasic monohydrate, Monosodium phosphate, Sodium dihydrogen phosphate monohydrate
Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-4 of 4
Compare
Product Number
Description
Pricing