
Secobarbital

Synonym(s):
5-(1-Methylbutyl)-5-(2-propenyl)-2,4,6(1H,3H,5H)-pyrimidinetrione, 5-Allyl-5-(1-methylbutyl)barbituric acid, Quinalbarbitone, Seconal
Empirical Formula (Hill Notation):
C12H18N2O3
CAS Number:
Molecular Weight:
238.28
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing