
Rhein

Synonym(s):
4,5-Dihydroxyanthraquinone-2-carboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthracenecarboxylic acid, Cassic acid, Chrysazin 3-carboxylic acid, Monorhein, Rheic acid, Rhubarb yellow
Empirical Formula (Hill Notation):
C15H8O6
CAS Number:
Molecular Weight:
284.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Research Area
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4