
(R)-(+)-α-Lipoic acid

Synonym(s):
(R)-(+)-1,2-Dithiolane-3-pentanoic acid, (R)-6,8-Dithiooctanoic acid, (R)-6,8-Thioctic acid
Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing