Skip to Content
BO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phthaldialdehyde
Synonym(s):
o
-Phthalaldehyde,
o
-Phthalic dicarboxaldehyde, Benzene-1,2-dicarboxaldehyde, OPA, Phthaldialdehyde
Linear Formula:
C
6
H
4
-1,2-(CHO)
2
CAS Number:
643-79-8
Molecular Weight:
134.13
EC Number:
211-402-2
Products
Product Category
building blocks (3)
aldehydes (1)
Brand
Millipore (1)
Sigma-Aldrich (4)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
P1378
Phthaldialdehyde,
≥97% (HPLC), powder or crystals
Expand
View Pricing
79760
Phthaldialdehyde,
suitable for fluorescence, ≥99.0% (HPLC)
Expand
View Pricing
8.21027
Phthaldialdehyde,
for synthesis
Expand
View Pricing
1.11452
Phthaldialdehyde,
for fluorometry
Expand
View Pricing
P0657
Phthaldialdehyde,
suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Expand
View Pricing