
Paromomycin sulfate salt

Empirical Formula (Hill Notation):
C23H45N5O14 · xH2SO4
CAS Number:
Molecular Weight:
615.63 (free base basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Quality Segment
Application
Mode of Action
Technique
Showing 1-3 of 3