Skip to Content
Merck
Palladium(II) acetylacetonate ≥99.9% trace metals basis

Palladium(II) acetylacetonate

Palladium(II) acetylacetonate ≥99.9% trace metals basis
Synonym(s):
Palladium(II) 2,4-pentanedionate, Pd(acac)2
Linear Formula:
Pd(C5H7O2)2
CAS Number:
Molecular Weight:
304.64

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing