
Orlistat Related Compound E

Synonym(s):
N-Formyl (S)-isoleucine (S)-1-[[(2S,3S)-3-hexyl-4-oxo-2-oxetanyl]methyl]dodecyl ester
Empirical Formula (Hill Notation):
C29H53NO5
CAS Number:
Molecular Weight:
495.73
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing