![O-[(N-Succinimidyl)succinyl-aminoethyl]-O′-methylpolyethylene glycol average Mn 750](/deepweb/assets/sigmaaldrich/product/structures/244/886/c80fd8d8-9a62-4a97-be17-32d83ffd1dfb/640/c80fd8d8-9a62-4a97-be17-32d83ffd1dfb.png)
O-[(N-Succinimidyl)succinyl-aminoethyl]-O′-methylpolyethylene glycol
![O-[(N-Succinimidyl)succinyl-aminoethyl]-O′-methylpolyethylene glycol average Mn 750](/deepweb/assets/sigmaaldrich/product/structures/244/886/c80fd8d8-9a62-4a97-be17-32d83ffd1dfb/640/c80fd8d8-9a62-4a97-be17-32d83ffd1dfb.png)
Synonym(s):
O-Methyl-O′-[(N-succinimidyl)succinyl-aminoethyl]polyethylene glycol 2′000, O-Methyl-O′-[2-(succinylamino)-ethyl]polyethylene glycol 2′000 N-succinimidyl ester, O-Methyl-O′-[(N-succinimidyl)succinyl-aminoethyl]polyethylene glycol, O-Methyl-O′-[2-(succinylamino)-ethyl]polyethylene glycol N-succinimidyl ester, Ome-PEG-NHS
CAS Number:
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-2 of 2