
N-Phenyl-p-phenylenediamine

Synonym(s):
4-Aminodiphenylamine, Variamine Blue RT Base
Linear Formula:
C6H5NHC6H4NH2
CAS Number:
Molecular Weight:
184.24
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing