Skip to Content
Merck
Methanol-13C,d4 99 atom % 13C, 99.5 atom % D

Methanol-13C,d4

Methanol-13C,d4 99 atom % 13C, 99.5 atom % D
Synonym(s):
Methyl alcohol-13C,d4, Methyl-13C,d3 alcohol-d
Linear Formula:
13CD3OD
CAS Number:
Molecular Weight:
37.06

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing