Skip to Content
Merck
Malachite green oxalate (C.I. 42000) for microscopy Certistain®

Malachite green oxalate (C.I. 42000)

Malachite green oxalate (C.I. 42000) for microscopy Certistain®
Synonym(s):
Malachite green oxalate (C.I. 42000)
Empirical Formula (Hill Notation):
C52H54N4O12
CAS Number:
Molecular Weight:
927.00

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2