
Lauric-d23 acid

Synonym(s):
Dodecanoic-d23 acid
Linear Formula:
CD3(CD2)10CO2H
CAS Number:
Molecular Weight:
223.46
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing