
Ioversol

Synonym(s):
Ioversol, N,N′-Bis(2,3-dihydroxypropyl)-5-[(hydroxyacetyl)(2-hydroxyethyl)amino]-2,4,6-triiodo-1,3-benzenedicarboxamide, N,N′-Bis(2,3-dihydroxypropyl)-5-[N-(2-hydroxyethyl)glycolamido]-2,4,6-triiodoisophthalamide
Empirical Formula (Hill Notation):
C18H24I3N3O9
CAS Number:
Molecular Weight:
807.11
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing