
Hyaluronic acid sodium salt from Streptococcus zooepidemicus

Synonym(s):
Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing