
Hexahydrocurcumin

Synonym(s):
(RS)-5-Hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)-3-heptanone
Empirical Formula (Hill Notation):
C21H26O6
CAS Number:
Molecular Weight:
374.43
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing