
Guanidine thiocyanate solution

Synonym(s):
Guanidine isothiocyanate, Guanidinium isothiocyanate, Guanidinium rhodanide, Guanidinium thiocyanate
Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Products
Product Category
Formula Weight
pH Value
Product Line
Showing 1-1 of 1
Compare
Product Number
Description
Pricing