
Gemfibrozil impurity E

Synonym(s):
(EZ)-2,2-Dimethyl-5-[2,5-dimethyl-4-(propene-1-yl)phenoxy]valeric acid
Empirical Formula (Hill Notation):
C18H26O3
CAS Number:
Molecular Weight:
290.40
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing