
Galactosyltransferase from bovine milk

Synonym(s):
Lactose Synthase, UDP-galactose:D-glucose 4-β-galactosyltransferase
CAS Number:
Products
Brand
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing