
Fura 2-AM

Synonym(s):
Fura-2 pentakis(acetoxymethyl) ester, FURA 2/AM
Empirical Formula (Hill Notation):
C44H47N3O24
CAS Number:
Molecular Weight:
1001.85
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Showing 1-3 of 3