
Frangulin B

Synonym(s):
3-(D-Apio-β-D-furanosyloxy)-1,8-dihydroxy-6-methyl-9,10-anthracenedione
Empirical Formula (Hill Notation):
C20H18O9
CAS Number:
Molecular Weight:
402.35
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing