
Form-2′,4′-xylidide

Synonym(s):
N-(2,4-Dimethylphenyl)formamide
Linear Formula:
(CH3)2C6H3NHCHO
CAS Number:
Molecular Weight:
149.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing