
Fluorene

Synonym(s):
Fluorene
Empirical Formula (Hill Notation):
C13H10
CAS Number:
Molecular Weight:
166.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing