
Fexofenadine Related Compound A

Synonym(s):
4-[4-[4-(Hydroxydiphenylmethyl)-1-piperidinyl]-1-oxobutyl]-α,α-dimethylphenylacetic acid
Empirical Formula (Hill Notation):
C32H37NO4
CAS Number:
Molecular Weight:
499.64
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2